SMS software compatibility with WINDOWS
Description:
This is articles explains the compatibility of SMS and its different features with the Windows versions.
SMS compatibility with Windows:
| SMS version | SQL version | Windows compatibility | "sa" SQL Administrator Account Removal | Vision compatibility | Comment |
| SMS 8.2.x | SQL SQL Express Server 2012/2019 | Win 10/ Win 11 | No | Yes | |
| SMS 8.3.x | SQL SQL Express Server 2022 | Win 11 | Yes, support secure login | No | Currently for Pc installed with OXSAS and SMS or without Vision |
Note that Vision software will not be compatible with WIN 11 before September 2026. This date is not commitment but an information.