Thermo Fisher Scientific
XOA Soft News 080-1 - Migrate from WinXRF UQ5 to OXSAS UQi
Description: 
OXSAS 2 introduced the Integrated UniQuant for new instruments only.
Migration from WinXRF 3.x + UniQuant 5 (UQ5) to OXSAS + Integrated UniQuant (UQi) is now possible on field instruments.
This note describes the steps to migrate data from WinXRF 3.x + UQ5 to OXSAS + UQi and describes the additional tasks to perform in OXSAS database regarding UQi for a fully usable system.
Environment:
ARL XRF instruments using WinXRF + UniQuant / OptiQuant v5 with 9 monitors