XOA Soft Note 039-4 - Installation Tips checklist and Windows versions
Description:
WinOE / XRF versions 3.y-z uses Windows® XP / 2000 components like IIS (Internet Information Services) and IE (Internet Explorer). Special care must be taken when configuring a computer and connecting it to an « administrated » network. The validated Windows versions compared to WinOE/XRF version are also a subject of concern.
Environment:
All ARL OE and ARLXRF instruments with WinOE/XRF v.3.x-y software
Attachment(s):
File | Last Modified |
---|---|
XO Soft Note 39-4 [Installation Tips] 1106.pdf | June 10, 2022 |
Soft WinOE_XRF [Installation Tips CheckList] 3-5.pdf | June 10, 2022 |