XOA Soft Note 064-1 - Problem during instrument configuration
Description:
During instrument initialization, a Sax Script error may be reported while the “XY_configuration.bas” script is sent to the ICS. That error prevents the further transmission of “hi…” configuration commands, causing problems during instrument analysis as some or all magazines positions are not defined.
This note is to explain the actions to be taken in order to correct the problem.
Environment:
ARL XRF instruments using X-Y or OMEGA magazine delivered with OXSAS up to version 1.1.3.644
Attachment(s):
File | Last Modified |
---|---|
XY_Configuration.zip | June 10, 2022 |
XR Soft Note 64-2 [XY configuration error] 0708.pdf | June 10, 2022 |