XOA Soft Note 078-1 - Analytical Assistant data update
Description:
Since OXSAS v. 2.1.2a, it is possible upgrading from OXSAS v. 1.x to v. 2.x (see Software Technical newsletter N° 34).
Since OXSAS 2.1.3c it’s now possible migrating from an OXSAS+UQ5 license to OXSAS with UQi (Integrated UniQuant) license.
This Technical Note explains the different steps to smoothly perform this upgrade on customer sites.
Environment:
All ARL instruments using OXSAS
Attachment(s):
File | Last Modified |
---|---|
BEA12802-XR Soft Note 78-1 [AA issues in OXSAS] 0514.zip | June 10, 2022 |