Thermo Fisher Scientific
XO Soft News 030-1 - UniQuant and OptiQuant Monitor Samples
Description: 
UniQuant, the well-known standard-less analysis software, is delivered with a box containing the software installation CD, the protection dongle and the Monitors samples used for Drift correction of the instrument (small picture below).
Up to now, the box contained 18 Monitors (small picture below). From now on, the UniQuant / OptiQuant box will contain 10 Monitors (big picture below) as 8 of the 18 are never used in field.
Environment:
All new ARL WD XRF instruments with UniQuant or OptiQuant option