UniQuant / OptiQuant Upgrade
Description:
The UniQuant / OptiQuant version upgrade kit is attached.
For more details on this topic or a more recent version please follow this link: UniQuant download.
Environment:
- UniQuant version 5 is for WD-XRF and version 6 and 7is for ED-XRF
- The integrated version of UniQuant, UQi, as a module of OXSAS is for WD-XRF only.
Attachment(s):
| File | Last Modified |
|---|---|
| BEA10241-Zuq556.exe | January 06, 2022 |