Thermo Fisher Scientific
CCU 1.2
Description: 
This document contains installers for the CCU version 1.2 valid for X900
Step 1: Open the SynopticView
Launch the SynopticView on your system.
Step 2: Navigate to Backup & Restore
From the top menu, go to:
Tools → Backup & Restore
Step 3: Import Firmware Data
  • Click on Import Data Entry.
  • Either:
    • Browse and select the firmware CCU installer file, or
    • Drag and drop the firmware installer into the import window.
  • Click OK to confirm the selection.
Step 4: Start Software Installation
Click on Start Software Installation to begin the firmware update process.
Step 5: Monitor Installation
Wait until the installation completes successfully. Do not close the tool or interrupt the process during installation.
⚠️ Important Note
This method is only valid for updating firmware from CCU version 1.1 to CCU version 1.2.
Environment:
ARL X900
  • Was this article helpful?