OXSAS 2.7.2.A Software Update
OXSAS Software Installation file for version 2.7.2.A together with SQL Server Express 2019 and installation guide released on 16/10/2023
Description:
Download OXSAS software with SQL Server Express 2019 for updating from prior version 2.4.1
Installer Setup_Oxsas_2.x.yyyy.exe is for updating from previously installed version 2.x
For updating OXSAS version 1.x please contact our Upgrade Team
Environment:
- OXSAS version 2.x (this version is for updating existing OXSAS version 2.x only)
- Windows 10 is supported since OXSAS 2.5.1
- SQL Server Express 2019 is supported for OXSAS 2.7.2.A
Attachment(s):
| File | Last Modified |
|---|---|
| 312486-A_EN [OXSAS 2.7.2A Release notes].pdf | October 17, 2023 |
| Setup_Oxsas_2.7.2.5836_a.zip | October 17, 2023 |