OXSAS 2.7.3.B Software Update
Description:
Download OXSAS software with SQL Server Express 2019 for updating from prior version 2.4.1
Installer Setup_Oxsas_2.x.yyyy.exe is for updating from previously installed version 2.x
For updating OXSAS version 1.x please contact our Upgrade Team
Environment:
- OXSAS version 2.x (this version is for updating existing OXSAS version 2.x only)
- Windows 10 is supported since OXSAS 2.5.1
- SQL Server Express 2019 is supported for OXSAS 2.7.2.A
Attachment(s):
| File | Last Modified |
|---|---|
| 313497-B_EN [OXSAS 2.7.3B Release notes].pdf | August 26, 2024 |
| OXSAS 2_7_3_53B.zip | August 26, 2024 |
| INSTALL_SQL2012.exe | August 20, 2024 |