Thermo Fisher Scientific
Oxsas 2.7.1.5620 b Software Update
Description: 
Download OXSAS software with SQL server 2012 for updating from prior version 2.4.1
Installer Setup_Oxsas_2.x.yyyy.exe is for updating from previously installed version 2.x
For updating OXSAS version 1.x please contact our Upgrade Team
Environment:
  • OXSAS version 2.x (this version is for updating existing OXSAS version 2.x only)
  • Windows 10 is supported since OXSAS 2.5.1
  • MSSQL 2012 is supported since OXSAS 2.4.1
  • Was this article helpful?