Thermo Fisher Scientific
GC Showing Not Ready Status
Issue
My GC does not reach the ready status.
Environment
Gas Chromatography
Resolution
  • Check the status displayed by the GC for any message.
  • Check if all gases are available, check pressures, flows and temperatures on the GC.
  • Check on GC status if all set points are reached (check the oven, injectors, detectors) With FID ,check if flame is on Verify the GC methods and the configuration, check the ready delay.
  • Check the GC handshaking parameters. 
  • Was this article helpful?