PS000012 - Product Specifications for the Thermo Scientific TRACE 1600 Series GC
The Thermo Scientific TRACE 1600 Series Gas (GC) maximizes uptime and profitability, offering interchangeable injector and detector modules for flexible configurations. The TRACE 1600 GC features a single-button interface, while the TRACE 1610 GC includes an advanced touch screen for enhanced user interaction. Both models provide reliability and ease of use, with features like the iConnect Column Lock, HeSaver-H2Safer technology, and integrated video guides for maintenance.
Description:
This document describes the Trace 1600 Series specifications
Environment:
Trace 1600 Series
Attachment(s):
| File | Last Modified |
|---|---|
| Product Specification - TRACE 1600 Series Gas Chromatograph.PDF | June 18, 2024 |