Thermo Fisher Scientific
FID Module Troubleshooting guide
To troubleshoot the Trace 1310 FID Module, ensure it is properly positioned, securely screwed, and running the latest firmware. Disconnect the column and close its inlet for better troubleshooting. Key issues include flame not igniting, no signal, high signal, high noise, spikes on the chromatogram, and detector not heating. Solutions involve checking gas connections, cleaning components, verifying electrical connections, and potentially replacing the module.
Description: 
This document describes how to troubleshoot a Trace 1300 / 1600 Series FID module.
Environment:
Trace 1300
Trace 1310
Trace 1600
Trace 1610
  • Was this article helpful?