Thermo Fisher Scientific
NPD Module Troubleshooting guide
Ensure the NPD module on the Trace 1300-1310 is positioned, secured, and updated. Disconnect the column and close its inlet for troubleshooting. Issues include no response, high signal, low response, FID-like response, negative baseline upsets, unstable baselines, excessive tailing, and non-heating. Solutions involve checking power, connections, gas calibrations, current values, and replacing components.
Description: 
This document describes how to troubleshoot a Trace 1300 / 1600 Series NPD Module
Environment:
Trace 1300
Trace 1310
Trace 1600
Trace 1610
  • Was this article helpful?