TRACE 1300 / 1600 Series Service Tool Manual - Rev A
The TS1300ST program is used for setting LAN communication parameters and updating the firmware on the GC. Installation involves inserting a provided DVD and following on-screen prompts. After installation, the program scans connected equipment, allowing users to set IP configurations and update firmware as needed. Only outdated firmware is updated. The program is closed using on-screen commands. Technical setup should be performed by Thermo Fisher Scientific personnel.
Description:
The TS1300ST program is used for setting the LAN Communication parameters, and for updating the Firmware version on the GC.
Environment:
Trace 1300 Series
Trace 1600 Series
Attachment(s):
| File | Last Modified |
|---|---|
| TRACE1300_1310_ServiceTools_Manual.pdf | June 19, 2024 |