Thermo Fisher Scientific
TRACE 1300 / 1600 Series Module Calibration Software V 1.03.04
The Trace 1300 Series Calibration Software is for Service Team module calibration using standard detector outfit, calibrated flowmeter, thermometer, and barometer. Install by running "setup.exe" and following prompts. Configure instruments by setting the GC IP address. Calibration involves selecting parameters, following on-screen instructions, and generating a report. Additional functions include setting oven temperature offsets and electronic cassette serial numbers.
Description: 
The modules calibration required the use of:
  • The standard outfit of the relative detector
  • A calibrated flowmeter
  • A calibrated thermometer (for oven temperature calibration)
  • A calibrated barometer (for inlets calibration)
Environment:
Trace 1300 Series
Trace 1600 Series
  • Was this article helpful?