Thermo Fisher Scientific
Configuring the Triplus RSH / 100 LS Autosampler to use 2 or 10 mL
Issue
Configuring the Triplus RSH / 100 LS Autosampler to use 2 or 10 mL - CV Magnetic Sample Vials for Headspace / SPME Analysis
Environment
  • Triplus RSH
  • Headspace
  • SPME analysis
Resolution
  1. Switch the Agitator’s Vial Type Position from 20-CV Magnetic to 10-CV Magnetic in the Extended User Level Administrator Mode of Virtual Terminal.
  2. Switch the Trayholder's Tray Vial Type from 20-CV Magnetic to 10-CV Magnetic in the Extended User Level Administrator Mode of Virtual Terminal.
  3. Install the 6 - 2 or 10 mL Agitator spacers into the 6 heated Agitator vial position spots.
The Spacer PNs:
  • TriPlus RSH - Agitator Adapter for 2mL Vial (Set of 6) P/N1R77010-1067
  • TriPlus RSH - Agitator Adapter for 10mL Vial (Set of 6) P/N1R77010-1068
  • Was this article helpful?