Part Number for 4 mL Wash Vial in AI/AS 1310 Gas Chromatography Autosampler
For the AI/AS 1310 Gas Chromatography Autosampler, the 4 mL wash vial part number is 24014031. It comes in a set of 125 screw top solvent vials and requires a screw top cap with septa, part number 60180-516. Ensure compatibility by cross-referencing with other parts
Issue:
Determining the part number for the 4 mL wash vial in the AI/AS 1310 Gas Chromatography Autosampler.
Environment:
- Product: AI/AS 1310 Gas Chromatography Autosampler
- Part Number: 24014031 - 4 mL Screw Top Solvent Vials, (set of 125)
- Document Reference: AI/AS 1310 Autosampler: Spare Parts Guide
Resolution:
- The 4 mL wash vial is listed under the description "4 mL Screw Top Solvent Vials, (set of 125)".
- The part number for the 4 mL wash vial is 24014031.
- This part requires a screw top cap with septa, part number 60180-516.
- Cross-reference with other parts to confirm compatibility.