Part Number for 4 mL Wash Vial in AI/AS 1610 Gas Chromatography Autosampler
For the AI/AS 1610 Gas Chromatography Autosampler, the 4 mL wash vial part number is 4-SVQ. It comes in a set of 500 screw top solvent vials and requires a screw top cap with septa, part number 6ASC9ST1. Ensure compatibility by cross-referencing with other parts
Issue:
Determining the part number for the 4 mL wash vial in the AI/AS 1610 Gas Chromatography Autosampler.
Environment:
- Product: AI/AS 1610 Gas Chromatography Autosampler
- Part Number: 4-SVQ - Vial, 4 mL clear screw glass (9 mm thread), Solvent vial for AI/AS 1310/1610, pkg. 500
- Document Reference: AI/AS 1610 Autosampler: Spare Parts Guide
Resolution:
- The 4 mL wash vial is listed under the description "Vial, 4 mL clear screw glass (9 mm thread), Solvent vial for AI/AS 1310/1610, pkg. 500".
- The part number for the 4 mL wash vial is 4-SVQ.
- This part requires a screw top cap with septa, part number 6ASC9ST1 which the desctiption is "Cap, Blue screw (9 mm) (PP: white silicone/red PTFE septa: 1.0 mm thickness) with AVCS technology, Performance Level 2, 100/Pack".
- Cross-reference with other parts to confirm compatibility.