Thermo Fisher Scientific
19050725 - Rev. A - LVI SSL Kit TRACE 1310
Install the Large Volume Splitless injector kit, which includes dummy filters, o-rings, a connector, a precolumn, splitless liners, and software. Ensure the GC is in standby, cool, and the carrier gas is off. Replace filters with dummy filters, install the precolumn, and connect it to the analytical column. Restore autosampler alignment, turn on the carrier gas, and set the working conditions. Increase oven temperature to 200°C for a good seal.
Description: 
The Large Volume Splitless injector is a setup of the standard splitless injector, where the introduction of large
amount of liquid samples can be performed manually, or with the TriPlus RSH or AI/AS 1310 autosampler.
Environment:
Trace 1300
Trace 1310
Trace 1600
Trace 1610
  • Was this article helpful?