31709671 - Rev. C - TriPlus 300 HS User Guide
The TriPlus 300 Headspace Sampling System User Guide details the operation, installation, and maintenance of the automated sampler for gas chromatographic analysis of volatile compounds in various matrices. The system includes a Main Unit with an oven and sampling system, and an Autosampler Unit for up to 120 samples. Safety precautions, installation steps, and optimization tips are provided to ensure accurate performance.
Description:
This manual describes the features and the components of the TriPlus 300 Headspace sampling system. Inside, you will find all the informations necessary for the routine operations of your sampling system. This includes operating procedures, sample injection techniques, diagrams, and the description of the major components.
Environment:
Triplus 300HS
Attachment(s):
| File | Last Modified |
|---|---|
| 31709671 (Rev. C) TriPlus 300 HS User Guide.pdf | June 19, 2024 |