31709711 - Rev A - TRACE 1300 series and TRACE 1600 Series Dual Detector Microfluidics Installation Guide
Install the Deans Switch Microfluidics Kit (P/N 19005580) on your TRACE 1300/1310 GC using the provided components and DVD. The DVD includes the Deans Switch Calculator program and the installation guide in PDF format. The kit allows splitting the eluent flow from the analytical column to two detectors. Ensure proper installation by following the detailed instructions, including connecting the tubing and using the provided tools.
Description:
Instructions for installing the Dual Detector Microfluidics.
Environment:
- Trace 1300
- Trace1310
- Trace 1600
- Trace 1610
- ISQ GC Series
- TSQ GC Series
Attachment(s):
| File | Last Modified |
|---|---|
| 31709741 - Rev A - TRACE 13001310 Deans Switch Microfluidics Kit.pdf | October 09, 2025 |
| 31709711 - Rev A - TRACE 1300 and TRACE 1310 Dual Detector Microfluidics Installation Guide.pdf | October 09, 2025 |