31709712 - Rev. D - Hot Injection Adapter Kit
Install the Hot Injection Adapter kit (PN 19050733) on the SSL/SSLBKF injector for gas sampling. Assemble the adapter, put the GC in standby mode, and cool the oven, injector, and detector. Power off the GC, remove the column, and take out the injector module. Install the new module flap cover, reinsert the injector, and remove the top parts. Replace the liner and seal, install the spacer plate, and reinstall the injector body. Attach the adapter, reconnect the column, and check for leaks.
Description:
This guide provides the instruction for installing the Hot Injection Adapter kit (P/N 19050733) on the
SSL/SSLBKF injector for the use with an external device for gas sampling.
Environment:
Trace 1300
Trace 1310
Trace 1600
Trace 1610
Attachment(s):
| File | Last Modified |
|---|---|
| 31709712 (Rev. D) Hot Injection Adapter Kit.pdf | August 26, 2025 |