31709721 -Rev. A - Pressure Regulator Kit GSV
Install the kit (P/N 19050763) to adjust the carrier gas pressure for the Gas Sampling Valve (GSV) injector if your carrier line cannot reach 450 kPa (68 psi; 4.5 bar). Connect the kit at a convenient point on the carrier gas line, such as near the gas cylinder or GC. The kit includes a pressure regulator, manometer, tee union, fittings, bracket, and 3-m copper tubing. Refer to the schematic diagrams for connection details.
Description:
This guide provides instructions for installing the kit P/N 19050763 for adjusting the pressure of the carrier gas to
correctly supply the Gas Sampling Valve (GSV) injector.
Environment:
Trace 1300
Trace 1310
Trace 1600
Trace 1610
Attachment(s):
| File | Last Modified |
|---|---|
| 31709721 (Rev. A) Pressure Regulator Kit GSV.pdf | June 18, 2024 |