Thermo Fisher Scientific
31709737 - Rev. D - Helium Saver Module User Guide
Install the Instant Connect Helium Saver Injector Module (P/N 19050763) on the TRACE 1300/1310 GC to reduce helium consumption. Assemble the module, connect helium and nitrogen supplies, and ensure proper gas purification. Follow instructions for setting carrier gas parameters, injection modes, and configuring split/splitless operations. Regularly maintain the module by replacing consumables like septa, liners, and filters. Optimize helium conservation by adjusting gas flows.
Description: 
This guide provides technical information and installation instructions for the Thermo Scientific™ Instant Connect Helium Saver Injector Module.
Environment:
Trace 1300
Trace 1310
Trace 1600
Trace 1610
  • Was this article helpful?