31709738 - Rev. E - On Column System User Guide
Install the On-Column Injector (OCI) system on the TRACE 1300/1310 GC, including the Auxiliary Gas Module and Aux Temperature/Cryo Module. Connect the gas lines and electrical connections, ensuring proper alignment and secure fittings. Configure the OCI module via the GC interface or Chromatography Data System (CDS), setting carrier gas parameters and secondary cooling timings. Perform a column leak check, calibrate the carrier gas flow, and condition the column. Replace panels and covers after
Description:
This guide provides the instructions for installing and operating with the On-Column injector (OCI) system for Thermo Scientific™ TRACE™ 1300 and TRACE™ 1310 Gas Chromatograph.
Environment:
Trace 1300
Trace 1310
Trace 1600
Trace 1610
Attachment(s):
| File | Last Modified |
|---|---|
| 31709738 (Rev. E) On_Column System_User_Guide.pdf | June 18, 2024 |