31709739 - Rev C - TRACE 1310 Auxiliary Oven: Valve Actuator Cooling Kit Installation Guide
Install the Valve Actuator Cooling Kit (PN 19071023) on the TRACE 1310 Auxiliary Oven to extend actuator lifetime at high temperatures (150°C to 270°C). For temperatures above 270°C, use high-temperature actuators without the cooling fans. Power off and disconnect the GC and Auxiliary Oven. Install the cooling fans on the L-shape cover, connect the electrical components, and reassemble the system. Ensure proper reconnection of all components and power on the system.
Description:
This guide provides instructions for installing the Valve Actuator Cooling Kit, PN 19071023 on your Thermo Scientific™ TRACE™ 1310 Auxiliary Oven equipped with rotary valves.
Environment:
Trace 1300
Trace 1310
Trace 1600
Trace 1610
Auxiliary Oven
Attachment(s):
| File | Last Modified |
|---|---|
| Valve Actuator (Rev. C) Cooling Kit IG.pdf | June 18, 2024 |