31709740 - Rev D - Deans Switch Microfluidics Installation Guide
Install the Deans Switch Microfluidics Kit (PN 19005580) on TRACE 1300/1310 GCs to apply analytical cuts on chromatograms. Install the Auxiliary Gas Module, mount the Deans Switch, and connect the tubing. Ensure all gas and electrical connections are secure before restarting the GC. Install the Deans Switch Calculator program, configure the system, and set analytical parameters. Follow the user guide for detailed instructions.
Description:
This guide provides instructions for installing the Deans Switch Microfluidics on your TRACE 1300 or TRACE 1310 GC.
Environment:
Trace 1300
Trace 1310
Trace 1600
Trace 1610
ISQ GC Series
TSQ GC Series
Attachment(s):
| File | Last Modified |
|---|---|
| 31709740 (Rev D) Deans Switch Microfluidics Installation Guide.pdf | June 18, 2024 |