31715015 - June 15 - Methanizer Assembly User Guide
Install the methanizer assembly in the TRACE 1310 Auxiliary Oven by removing the protective grid and inserting the methanizer into the slots. Connect the necessary wires and components. Maintain the methanizer by replacing the catalyst when exhausted and performing leak tests. Activate the catalyst by following specific temperature and hydrogen flow protocols. Troubleshoot issues like no peaks or absent CO/CO2 peaks by checking connections and replacing components as needed.
Description:
This note provides instruction for installing, maintaining, and troubleshooting the methanizer assembly for the TRACE 1310 Auxiliary Oven.
Environment:
Trace 1300 Series
Trace 1600 Series
Attachment(s):
| File | Last Modified |
|---|---|
| Methanizer Assembly User Guide PN 31715015_June 15.pdf | June 18, 2024 |