Thermo Fisher Scientific
31715016 - Rev. B - Split Fitting Kit
Install the split option kit P/N 19071018 in the TRACE 1310 Auxiliary Oven by powering off the GC and removing the L-shape covers to access the interior. Modify the gas inlet port for controlling split flow by plugging the gas inlet fitting and replacing the M5 cap with an M5 gas inlet fitting. Position the splitter and flow restrictor inside the oven according to the pneumatic circuit. Ensure flow restrictor is installed inside the auxiliary oven at a regulated temperature for flow stability.
Description: 
This note provides installing the kit P/N 19071018 for the use of one channel of the auxiliary gas module for controlling a split flow.
Environment:
Trace 1300
Trace 1310
Trace 1600
Trace 1610
  • Was this article helpful?