31717000 - Rev. A - NoVent Quick Reference Card
Install the No Vent™ Microfluidics kit on your TRACE 1300/TRACE 1310 GC by following the video and printable instructions on the provided USB stick. The kit includes a mounting bracket, deactivated tubing, No Vent Microfluidics Tee, SilTite™ FingerTite nuts and ferrules, SilFlow™ nuts and ferrules, and pre-swage tools. Access the video tutorial on your PC or TRACE 1310 touchscreen to ensure proper installation. The video does not interfere with GC operation.
Description:
NoVent Quick Reference Card
Environment:
Trace 1300
Trace 1310
Trace 1600
Trace 1610
Attachment(s):
| File | Last Modified |
|---|---|
| 31717000 (Rev. A) NoVent Quick Reference Card.pdf | June 18, 2024 |