MI-317000-0025 - Rev. A - Helium Saver H2 Safer User Guide
Install and maintain the Thermo Scientific Instant Connect Helium Saver-H2 Safer option for TRACE 1600/1610 GCs to reduce helium consumption and ensure safe hydrogen use. Follow the user guide for technical information, installation, and maintenance procedures. The kit includes components and instructions for operation modes, gas requirements, and consumables. Configure and plumb the system, check for leaks, and maintain components like septa, liners, and filters to ensure optimal performance.
Description:
This document provides technical information, installation instructions, and maintenance procedures for the Thermo Scientific™ Instant Connect Helium Saver-H2 Safer option.
Environment:
Trace 1600
Trace 1610
Attachment(s):
| File | Last Modified |
|---|---|
| MI-317000-0025 (Rev. A) Helium Saver H2 Safer User Guide.pdf | October 09, 2025 |