Thermo Fisher Scientific
MI-317000-0032 (Rev. C) TRACE 1600 Series PreInstallation Guide
Prepare your site for the installation of the Thermo Scientific TRACE 1600/1610 gas chromatograph by ensuring adequate entrance, workbench space, lighting, power, LAN network, and environmental conditions. Follow guidelines for handling gases and cryogenic cooling. Inspect and protect the shipment upon arrival, and complete the Installation Request Form. Thermo Fisher Scientific's Field Service Engineer will install the system and confirm performance tests.
Description: 
This guide contains detailed information for preparing your site for installing a Thermo Scientific™ TRACE 1600™ GC or TRACE™ 1610 GC.
Environment:
Trace 1600
Trace 1610
  • Was this article helpful?