AI - AS Routine Service Intervals
Topic
Here are some maintenance procedures that should be performed routinely.
Environment
AI/AS Autosampler
Steps/Procedure
- Disposing of contents in the waste vial.
- Changing wash solvents approximately every quarter (depending on run frequency).
- Automated cleaning of the syringe. Approximately after every 100 injections.
- Clean the plunger after every 1000 injections or as needed.
Additional Information
- The AI/AS autosampler hardly requires any maintenance.
- Most of the maintenance procedures can be completed in 5 minutes or less.
- The common issues that typically occur are related to contamination in the wash vials, and syringe contamination or damage.