How to Run Positive and Negative Autotune
Topic
How to Perform Positive and Negative Autotune on an ISQ EC or ISQ EM
Environment
- ISQ EC Mass Spectrometer
- ISQ EM Mass Spectrometer
- Chromeleon software
- HESI source
- Positive and Negative ESI tuning
Steps/Procedure
-
Complete the calibrant delivery and tuning preparation procedures.
-
From the detector ePanel, click Tune.
-
Locate the Positive and Negative Autotune controls.
Run Positive Autotune
-
Click ESI Positive Tune.
-
Monitor the tuning status and progress displayed on the tuning page.
-
Allow Positive Autotune to complete.
-
Open the generated positive tune report.
-
Confirm that the tune passed.
Run Negative Autotune
-
Return to the Tune screen.
-
Click ESI Negative Tune.
-
Monitor the displayed tuning progress.
-
Allow Negative Autotune to complete.
-
Open the generated negative tune report.
-
Confirm that the tune passed.
Review Tune Results
-
Locate Available Tune Results on the Tune screen.
-
Review the reports identified by their date and time.
-
Identify the positive tune report by the PESI designation.
-
Identify the negative tune report by the NESI designation.
-
Confirm that both tuning programs passed before operating the detector.
Additional Information
- Both Positive and Negative Autotune must be performed even when the application uses only one polarity.
- Always run Positive Autotune before Negative Autotune.
- The training specifies retuning before and after repairs, before and after optics cleaning, or when known masses shift in the spectrum.
- If either tuning program fails, use ESI Diagnostic-Only Tune to begin troubleshooting.
- Additional troubleshooting information is available in the ISQ EC/EM Operator Manual and the ISQ EC and ISQ EM Single Quadrupole Mass Spectrometer: Routine Diagnostics and Troubleshooting training.