Thermo Fisher Scientific
ITEX Sampling on the TriPlus RSH SMART
Topic
In-Tube Extraction (ITEX) is a syringe-based headspace enrichment technique that uses a gas-tight syringe and a sorbent-filled micro-trap inside the syringe needle to trap and concentrate volatile compounds.
Environment
  • TriPlus RSH SMART autosampler
  • GC or GC-MS
  • ITEX syringe
  • Gas-tight syringe
  • Sorbent-filled micro-trap
  • Incubator
  • Nitrogen supply for trap cleaning
Steps/Procedure
  1. Place the sample in the incubator.
  2. Pre-equilibrate the sample at the selected constant temperature and agitation speed.
  3. Collect volatile compounds from the vial headspace using the ITEX syringe.
  4. Perform repeated syringe strokes through the headspace as defined by the method.
  5. Allow volatile analytes to become trapped and concentrated on the sorbent in the micro-trap.
  6. Insert the ITEX needle into the GC injector.
  7. Thermally desorb the trapped analytes through the GC inlet and into the analytical column.
  8. After analysis, heat the ITEX syringe and needle.
  9. Purge the syringe and needle with nitrogen to clean the trap.
Additional Information
The training identifies the following benefits of ITEX:
  • High sorbent capacity
  • Increased sensitivity at lower detection limits
  • Robustness
  • Ease of use
  • Long trap lifetime
The training references an ITEX-DHS GC-MS application for trace-level odorants in water, including isoborneol, 2-methyl-isoborneol, 2,4,6-trichloroanisole, and geosmin.
Source terminology note: The training materials are not completely consistent in how ITEX is categorized. One slide describes ITEX as a “syringe-based static headspace enrichment technique,” while the summary describes it as a “dynamic headspace enrichment technique.” This article therefore uses the broader source-supported description “syringe-based headspace enrichment technique” rather than resolving the discrepancy without additional technical documentation.