PTV Inlet Septum and Liner Service
Topic
PTV—Septum and Liner Change
Environment
- Trace GC 1300
- Trace GC 1600
Steps/Procedure
- Open the injector module flap.
- Unscrew the septum cap.
- Take out the septa using tweezers.
- Using the slotted stubby driver provided, unscrew and remove the liner cap.
- Use tweezers to remove the liner cap.
- Place the liner removal tool on-top of the liner and push to engage the liner.
- PTV glass liners are especially fragile, hence gently pull the liner out of the inlet.
- Insert a new liner with the white O-ring liner seal inserted.
- Gently push down the liner until it bottoms out and properly seated.
- Re-install the liner cap and screw on using the slotted stubby driver.
- Insert the new piece of septa.
- Screw on the septum cap and close the PTV module flap.
Additional Information
- Make sure to adhere to the safety rules as indicated in the Trace 1300 and 1600 user manuals.
- Cross-reference: KCS 024 — Trace 1610 Maintenance Status Counter Reset.