SSL Inlet Septum and Liner Service
Topic
SSL—Septum and Liner Change
Environment
Trace GC 1300 & 1600 Series
Steps/Procedure
- Open the injector module flap.
- Unscrew the septum cap.
- Take out the septa using tweezers.
- Unscrew the ring nut.
- Remove the septum holder/liner cap from the injector body head.
- Using a tweezer, take out the liner from the injector.
- Insert a new liner with the white O-ring seal as seen here.
- Re-install the septum holder/liner cap.
- Screw on the ring nut.
- Insert a new piece of septa.
- Screw on the septum cap.
- Close the module flap.
Additional Information
- Make sure to adhere to the safety rules as indicated in the Trace 1300 and 1600 user manuals.
- Cross-reference: KCS 024 — Trace 1610 Maintenance Status Counter Reset.