Trace 1610 Maintenance Status Counter Reset
Topic
There is a heart-shaped icon on the screen that provides alerts on maintenance status for the GC.
Environment
Trace 1610 GC
Steps/Procedure
- Click on each alert and reset the counters after maintenance.
- After the counters have been reset, the heart-shaped icon turns green with a check mark.
- This indicates that the GC is up to date with its maintenance schedule.
Additional Information
- This icon indicates the system maintenance status.
- By clicking on the icon, one can reset the counter after each maintenance step.
- For items replaced or maintained on the Trace 1610 GC, reset the counters to indicate working conditions.