Thermo Fisher Scientific
Trace GC Preventive Maintenance Intervals
Topic
Here is the maintenance schedule for the Trace GC.
Environment
  • Trace GC 1300 Series
  • Trace GC 1600 Series
Steps/Procedure
  • Clean exterior surfaces using lint-free cloth and/or soapy water — Quarterly or as needed.
  • Leak checks — Weekly.
  • Helium filters or gas scrubbers — Checked quarterly and replace yearly.
  • Injector maintenance:
    • Change Septa.
    • Change Liner.
    • Replace seals/O-rings.
    • Injection maintenance is sample dependent. Dirty matrices might require more inlet cleaning.
    • After every 100 injections.
    • Depending on instrument performance. Carryover and syringe contamination could contribute to a dirty liner.
    • Typically done when there are leaks.
    • Always utilize the leak check function after column installation or trimming.
  • Column maintenance:
    • Columns can last a long-time if properly taken care of.
    • Consider using guard columns or backflush connections to minimize downtimes.
    • Check for leaks using a leak detector or through the mass spectrometer (if connected).
    • Do this on a weekly basis.
Additional Information
  • The GC oven itself requires little to no maintenance.
  • Injectors or detectors require the most maintenance.
  • This e-learning is going to focus on just injector maintenance due to the coupling of the GC to the ISQ.
  • Column maintenance is another critical routine maintenance procedure.
  • Column maintenance is determined when instrument performance deteriorates, and troubleshooting indicates that a replacement or trimming is required.