19050725 - Rev. A - LVI SSL Kit TRACE 1310
Description:
The Large Volume Splitless injector is a setup of the standard splitless injector, where the introduction of large
amount of liquid samples can be performed manually, or with the TriPlus RSH or AI/AS 1310 autosampler.
Environment:
Trace 1300
Trace 1310
Trace 1600
Trace 1610
Attachment(s):
File | Last Modified |
---|---|
19050725 (Rev. A) LVI SSL Kit TRACE 1310.pdf | June 18, 2024 |