TRACE 1300 / 1600 Series Module Calibration Software V 1.03.04
Description:
The modules calibration required the use of:
- The standard outfit of the relative detector
- A calibrated flowmeter
- A calibrated thermometer (for oven temperature calibration)
- A calibrated barometer (for inlets calibration)
Environment:
Trace 1300 Series
Trace 1600 Series
Attachment(s):
File | Last Modified |
---|---|
TRACE 1300 SERIES CALIBRATION TOOL INSTALLATION AND USER MANUAL.pdf | June 17, 2024 |
Module_Calibration_SW_1.03.04.zip | June 17, 2024 |