PSB-IC-SYS-1014 - AS-AP Autosampler Needle Alignment
When performing needle alignment on an AS-AP autosampler, it must be configured in Push mode in Chromeleon Server Configuration to recognize the injection port properly. After alignment, it can be set back to Pull mode if needed. This will not affect the alignment. Contact Ion Chromatography Technical Support if issues persist.
Description:
When performing the needle alignment on an AS-AP autosampler, the needle must have access to the injection port, wash port and the two alignment marks on the vial tray in the red position.
Environment:
- AS-AP Autosampler Needle
- All SN
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-IC-SYS-1014 - AS-AP Autosampler Needle Alignment.PDF | January 07, 2022 |