PSB-IC-SYS-1015 - AS-AP Autosampler Needle Spring Assembly
Starting December 2011, the AS-AP autosampler needle spring assembly was upgraded with a stronger spring to address needle obstruction errors. If issues persist after eliminating common causes, replace the needle spring and end cap with new parts. Perform a needle force calibration in Chromeleon after installation. Contact Ion Chromatography Technical Support if needed.
Description:
Beginning in December of 2011, the needle spring assembly of the AS-AP autosampler has been upgraded with a stronger spring. This will allow the AS-AP to apply greater force when piercing a vial septum. This may address certain needle obstruction errors. This PSB details the field upgrade procedure for installing the stronger spring.
Environment:
- AS-AP Autosampler Needle Spring Assembly
- SN - All below 1112xxxx
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-IC-SYS-1015 - AS-AP Autosampler Needle Spring Assembly.PDF | January 07, 2022 |