PSB-IC-SYS-1017 - AS-AP Autosampler Carousel Vial Tray Doors
AS-AP carousel vial tray doors may be difficult to close due to friction, causing the needle carriage magnet to disengage and the door to remain open. To resolve this, adjust the carousel tray tub as described. If the issue persists, send the AS-AP unit for factory repair. Contact Ion Chromatography Technical Support if needed.
Description:
AS-AP carousel vial tray doors that are difficult to close due to friction between carousel door and the tray tub. The increased force needed to close the door can result in the disengagement of the needle carriage magnet, causing the door to remain open. This PSB describes the field repair steps for readjustment of the AS-AP carousel vial tray door.
Environment:
- AS-AP Autosampler Carousel Vial Tray Doors
- SN - All AS-AP units
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-IC-SYS-1017 - AS-AP Autosampler Carousel Vial Tray Doors.PDF | January 07, 2022 |