PSB-ICSP-1069 - AS-AP Autosampler Needle Port Seal Leakage
A new AS-AP needle holder was implemented in October 2016, requiring the NeedleForceAdjustmentDepth value to be updated to 2784 in the EEPROM. Units shipped between October 2016 and January 2017 were not updated, causing leaks. Corrective action involves adjusting the NeedleForceAdjustmentDepth to 2784 and recalibrating the needle force. Units made after January 2017 are updated. Older units need adjustments when replacing the XZ drive assembly.
Description:
A new version of the AS-AP needle holder in the XZ drive assembly has been implemented since October 2016. With this change, the default value 3800 in the property for NeedleForceAdjustmentDepth has been updated to 2784, and this value is stored in EEPROM of the AS-AP main board.
Environment:
- AS-AP Autosampler Needle holder
- SN < 17010623
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-ICSP-1069 - AS-AP Autosampler Needle Port Seal Leakage.PDF | January 07, 2022 |