PSB-ICSP-1158 - AS-AP Washport Assembly Kit
Thermo Fisher Scientific introduced consolidated part number B5100295, for the AS-AP Washport Assembly Kit. This kit includes all parts for replacing the complete wash port assembly, streamlining the process for Field Service Engineers. The kit is now available for order, ensuring efficient replacement with all required components. For specific repairs requiring a complete wash port assembly replacement, order P/N B51002935 and refer to the AS-AP Service manual for alignment procedures.
Description:
To streamline the process for Field Service Engineers when changing the complete wash port assembly, including the wash port, needle seat parts, and other hardware components, we have introduced a new consolidated part number: P/N B51002935 (KIT,ASSY,WASHPORT, AS-AP). This kit includes all necessary parts for replacing the entire wash port assembly. The new part number is now active and available for order in service ensuring that Field Service Engineers can efficiently carry out the complete wash port assembly replacement with all required components readily available.
Environment:
AS-AP Autosampler
Attachment(s):
| File | Last Modified |
|---|---|
| PSB-ICSP-1158-AS-AP Washport Assembly Kit.pdf | February 13, 2025 |