AS-AP Firmware v 1.9.0 and Release Notes
A new firmware version 1.9.0 for the AS-AP Autosampler was released on May 26, 2017, superseding version 1.8.0. Bug fixes include increased precision tolerances for position checking to reduce "Alignment not precise" and "End Point not reached" errors. Customers using 384 position well plates should not upgrade. Update the firmware via the Chromeleon Device Driver.
Description:
AS-AP Firmware version 1.9.0
Release Date 10 May 2017
Environment:
AS-AP Autosampler
Attachment(s):
| File | Last Modified |
|---|---|
| AS-AP Firmware 1.9.0.zip | November 21, 2021 |
| AS-AP Firmware 1.9.0 Release Notes.pdf | November 21, 2021 |