Thermo Fisher Scientific
AS-AP Autosampler Leaking Issue - Chinese
Description: 
This basic troubleshooting video identifies the possible components that cause the leakage. This video also identifies the leak sensor and the leak alarm on the AS-AP front light panel. After watching this video, operators will be able to remove the needle from the needle arm, manually check the broken needle, replace the broken needle seat, and identify the high backpressure injection valve.
Environment:
  • AS-AP Autosampler
  • Ion Chromatography
  • Was this article helpful?